4-(Aminomethyl)benzonitril Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-(Aminomethyl)benzonitrile HCl, α-Amino-p-tolunitrile Hydrochloride, 4-(Aminomethyl)benzonitrile Hydrochloride, 4-Cyanobenzylamine Hydrochloride

M-Clarity™ Qualitätsstufe = MQ100

631396-25G 631396-5G
SIAL631396-25GEA 99.3 EUR
SIAL631396-25G SIAL631396-5G
4-(Aminomethyl)benzonitril Hydrochlorid, Sigma-Aldrich®
4-(Aminomethyl)benzonitril Hydrochlorid
Formel: C₈H₉ClN₂
Molecular Weight: 168.62 g/mol
MDL: MFCD01861472
CAS-Nummer: 15996-76-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR