5-Acetyl-2-fluorphenylboronsäure

Lieferant: FLUOROCHEM

Synonyme: (5-Acetyl-2-fluorophenyl)boronic acid

016211-1G 016211-5G
FLUO016211-1GEA 76.2 EUR
FLUO016211-1G FLUO016211-5G
5-Acetyl-2-fluorphenylboronsäure
5-Acetyl-2-fluorphenylboronsäure
Formel: C₈H₈BFO₃
Molecular Weight: 181.9567 g/mol
MDL: MFCD06656263
CAS-Nummer: 870777-29-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR