4-(1-(Hydroxymethyl)cyclopropyl)phenylboronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4-(1-(Hydroxymethyl)cyclopropyl)phenyl)boronic acid

BB-5947-1G BB-5947-250M
COBBBB-5947-1GEA 1090 EUR
COBBBB-5947-1G COBBBB-5947-250M
4-(1-(Hydroxymethyl)cyclopropyl)phenylboronsäure
4-(1-(Hydroxymethyl)cyclopropyl)phenylboronsäure
Formel: C₁₀H₁₃BO₃
Molecular Weight: 192,0194 g/mol
MDL: MFCD12546571
CAS-Nummer: 1217501-10-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR