4'-Acetylbiphenyl-4-boronsäure

Lieferant: COMBI-BLOCKS

Synonyme: (4'-Acetyl-[1,1'-biphenyl]-4-yl)boronic acid

BB-6173-5G BB-6173-1G BB-6173-250M
COBBBB-6173-5GEA 2040 EUR
COBBBB-6173-5G COBBBB-6173-1G COBBBB-6173-250M
4'-Acetylbiphenyl-4-boronsäure
4'-Acetylbiphenyl-4-boronsäure
Formel: C₁₄H₁₃BO₃
Molecular Weight: 240,0600 g/mol
MDL: MFCD16295057
CAS-Nummer: 1029438-14-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR