4-Brom-4'-nitrobiphenyl

Lieferant: COMBI-BLOCKS

Synonyme: 4-Bromo-4'-nitro-1,1'-biphenyl

AN-1509-1G AN-1509-5G
COBBAN-1509-1GEA 265 EUR
COBBAN-1509-1G COBBAN-1509-5G
4-Brom-4'-nitrobiphenyl
4-Brom-4'-nitrobiphenyl
Formel: C₁₂H₈BrNO₂
Molecular Weight: 278,1014 g/mol
MDL: MFCD00024697
CAS-Nummer: 6242-98-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR