4-Cyan-4'-octyloxybiphenyl

Lieferant: CHEMPUR

Synonyme: 4'-Octyloxy-[1,1'-biphenyl]-4-carbonitrile

31-0006.500 31-0006.1
CHMP31-0006.500EA 0 EUR
CHMP31-0006.500 CHMP31-0006.1
4-Cyan-4'-octyloxybiphenyl
4-Cyan-4'-octyloxybiphenyl
Formel: C₂₁H₂₅NO
Molecular Weight: 307.44 g/mol
MDL: MFCD00075145
CAS-Nummer: 52364-73-5
EINECS: 257-878-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR