4-Cyan-4'-octyloxybiphenyl, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4'-Octyloxy-[1,1'-biphenyl]-4-carbonitrile

M-Clarity™ Qualitätsstufe = MQ100

338672-1G 338672-250MG
SIAL338672-1GEA 103 EUR
SIAL338672-1G SIAL338672-250MG
4-Cyan-4'-octyloxybiphenyl, Sigma-Aldrich®
4-Cyan-4'-octyloxybiphenyl
Formel: C₂₁H₂₅NO
Molecular Weight: 307.44 g/mol
MDL: MFCD00075145
CAS-Nummer: 52364-73-5
EINECS: 257-878-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR