4-Cyan-4'-pentyloxybiphenyl ≥99%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
4-Amyloxy-4'-cyanobiphenyl, 4-Cyano-4'-n-pentyloxybiphenyl, 5OCB, 4'-Amyloxy-4-biphenylcarbonitrile, 4'-Pentyloxy-4-biphenylcarbonitrile, 4'-Pentyloxy-[1,1'-biphenyl]-4-carbonitrile
B22477.03
B22477.06
B22477.03EA
52.3
EUR
B22477.03
B22477.06
4-Cyan-4'-pentyloxybiphenyl ≥99%
4-Cyan-4'-pentyloxybiphenyl
Purity: 99%
CAS No.: 52364-71-3
Molecular Formula: C18H19NO
Formula Weight: 281.42
Flash Point:
Formel:
C₁₈H₁₉NO Molecular Weight: 265,35 g/mol Schmelzpunkt: 48 °C Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00074879 CAS-Nummer: 52364-71-3 EINECS: 257-875-9 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...