Methyl-8-brom-6-chlor-4-hydroxychinolin-2-carboxylat

Lieferant: COMBI-BLOCKS

HC-2540-5G HC-2540-1G
COBBHC-2540-5GEA 303 EUR
COBBHC-2540-5G COBBHC-2540-1G
Methyl-8-brom-6-chlor-4-hydroxychinolin-2-carboxylat
Methyl-8-brom-6-chlor-4-hydroxychinolin-2-carboxylat
Formel: C₁₁H₇BrClNO₃
MDL: MFCD11855927
CAS-Nummer: 1133116-01-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR