Ethyl-6-brom-4-hydroxychinolin-3-carboxylat

Lieferant: COMBI-BLOCKS

Synonyme: Ethyl 6-bromo-4-hydroxyquinoline-3-carboxylate

HC-2328-5G HC-2328-1G
COBBHC-2328-5GEA 0 EUR
COBBHC-2328-5G COBBHC-2328-1G
Ethyl-6-brom-4-hydroxychinolin-3-carboxylat
Ethyl-6-brom-4-hydroxychinolin-3-carboxylat
Formel: C₁₂H₁₀BrNO₃
Molecular Weight: 296.1200 g/mol
MDL: MFCD00173362
CAS-Nummer: 122794-99-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR