Methyl-6-chinolincarboxylat

Lieferant: Thermo Scientific

KM10245DA KM10245EA
MAYBKM10245DAEA 0 EUR
MAYBKM10245DA MAYBKM10245EA
Methyl-6-chinolincarboxylat
Methyl-6-chinolincarboxylat
Formel: C₁₁H₉NO₂
Molecular Weight: 187,2000 g/mol
MDL: MFCD00956766
CAS-Nummer: 38896-30-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR