Ethyl-4-hydroxy-8-(trifluormethoxy)chinolin-3-carboxylat

Lieferant: Thermo Scientific

Synonyme: Ethyl 4-hydroxy-8-(trifluoromethoxy)quinoline-3-carboxylate

TL00807.1 TL00807.10 TL00807.50
MAYBTL00807.1EA 0 EUR
MAYBTL00807.1 MAYBTL00807.10 MAYBTL00807.50
Ethyl-4-hydroxy-8-(trifluormethoxy)chinolin-3-carboxylat
Ethyl-4-hydroxy-8-(trifluormethoxy)chinolin-3-carboxylat
Formel: C₁₃H₁₀F₃NO₄
Molecular Weight: 301,2180 g/mol
MDL: MFCD00221480
CAS-Nummer: 175278-35-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR