Ethyl-6-fluor-4-hydroxychinolin-3-carboxylat

Lieferant: COMBI-BLOCKS

Synonyme: Ethyl 6-fluoro-4-hydroxyquinoline-3-carboxylate

PY-7380-1G PY-7380-5G
COBBPY-7380-1GEA 63.4 EUR
COBBPY-7380-1G COBBPY-7380-5G
Ethyl-6-fluor-4-hydroxychinolin-3-carboxylat
Ethyl-6-fluor-4-hydroxychinolin-3-carboxylat
Formel: C₁₂H₁₀FNO₃
Molecular Weight: 235,22 g/mol
MDL: MFCD00173345
CAS-Nummer: 318-35-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR