Methyl-4-chlor-8-(trifluormethyl)chinolin-2-carboxylat
Lieferant: COMBI-BLOCKS
HC-2288-5G
HC-2288-1G
COBBHC-2288-5GEA
0
EUR
COBBHC-2288-5G
COBBHC-2288-1G
Methyl-4-chlor-8-(trifluormethyl)chinolin-2-carboxylat
Methyl-4-chlor-8-(trifluormethyl)chinolin-2-carboxylat
Formel:
C₁₂H₇ClF₃NO₂ |
MDL:
MFCD11855877 CAS-Nummer: 1133115-62-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...