Methyl-4-hydroxy-6-methylchinolin-2-carboxylat

Lieferant: COMBI-BLOCKS

HC-6981-5G HC-6981-1G
COBBHC-6981-5GEA 451 EUR
COBBHC-6981-5G COBBHC-6981-1G
Methyl-4-hydroxy-6-methylchinolin-2-carboxylat
Methyl-4-hydroxy-6-methylchinolin-2-carboxylat
Formel: C₁₂H₁₁NO₃
MDL: MFCD11504934
CAS-Nummer: 950265-42-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR