Methyl-4-hydroxy-8-methylchinolin-2-carboxylat

Lieferant: COMBI-BLOCKS

HC-2764-1G HC-2764-5G
COBBHC-2764-1GEA 98.3 EUR
COBBHC-2764-1G COBBHC-2764-5G
Methyl-4-hydroxy-8-methylchinolin-2-carboxylat
Methyl-4-hydroxy-8-methylchinolin-2-carboxylat
Formel: C₁₂H₁₁NO₃
MDL: MFCD12756432
CAS-Nummer: 849022-03-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR