Ethyl-4-hydroxy-8-methylchinolin-3-carboxylat

Lieferant: COMBI-BLOCKS

Synonyme: Ethyl 8-methyl-4-oxo-1,4-dihydroquinoline-3-carboxylate

PY-7939-1G PY-7939-5G
COBBPY-7939-1GEA 141 EUR
COBBPY-7939-1G COBBPY-7939-5G
Ethyl-4-hydroxy-8-methylchinolin-3-carboxylat
Ethyl-4-hydroxy-8-methylchinolin-3-carboxylat
Formel: C₁₃H₁₃NO₃
Molecular Weight: 231,2500 g/mol
MDL: MFCD00173390
CAS-Nummer: 77156-75-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR