Methyl-4-hydroxy-6-(trifluormethyl)chinolin-2-carboxylat

Lieferant: COMBI-BLOCKS

HC-2537-1G HC-2537-250M
COBBHC-2537-1GEA 420 EUR
COBBHC-2537-1G COBBHC-2537-250M
Methyl-4-hydroxy-6-(trifluormethyl)chinolin-2-carboxylat
Methyl-4-hydroxy-6-(trifluormethyl)chinolin-2-carboxylat
Formel: C₁₂H₈F₃NO₃
MDL: MFCD00278339
CAS-Nummer: 123158-31-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR