Ethyl-4-hydroxy-6-(trifluormethyl)chinolin-3-carboxylat

Lieferant: COMBI-BLOCKS

Synonyme: Ethyl 4-hydroxy-6-(trifluoromethyl)quinoline-3-carboxylate

HC-2304-1G HC-2304-25G HC-2304-5G
COBBHC-2304-1GEA 0 EUR
COBBHC-2304-1G COBBHC-2304-25G COBBHC-2304-5G
Ethyl-4-hydroxy-6-(trifluormethyl)chinolin-3-carboxylat
Ethyl-4-hydroxy-6-(trifluormethyl)chinolin-3-carboxylat
Formel: C₁₃H₁₀F₃NO₃
MDL: MFCD00173369
CAS-Nummer: 26893-12-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR