Methyl-4-hydroxy-7-(trifluormethyl)chinolin-2-carboxylat
Lieferant: COMBI-BLOCKS
Synonyme:
Methyl 4-hydroxy-7-(trifluoromethyl)quinoline-2-carboxylate
HC-2226-5G
HC-2226-1G
COBBHC-2226-5GEA
884
EUR
COBBHC-2226-5G
COBBHC-2226-1G
Methyl-4-hydroxy-7-(trifluormethyl)chinolin-2-carboxylat
Methyl-4-hydroxy-7-(trifluormethyl)chinolin-2-carboxylat
Formel:
C₁₂H₈F₃NO₃ Molecular Weight: 271.1920 g/mol |
MDL:
MFCD11504916 CAS-Nummer: 1072944-69-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...