Methyl-4-hydroxy-8-(trifluormethyl)chinolin-2-carboxylat

Lieferant: COMBI-BLOCKS

HC-6948-5G HC-6948-1G
COBBHC-6948-5GEA 451 EUR
COBBHC-6948-5G COBBHC-6948-1G
Methyl-4-hydroxy-8-(trifluormethyl)chinolin-2-carboxylat
Methyl-4-hydroxy-8-(trifluormethyl)chinolin-2-carboxylat
Formel: C₁₂H₈F₃NO₃
MDL: MFCD11053782
CAS-Nummer: 1065074-52-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR