Ethyl-4-hydroxy-8-(trifluormethyl)chinolin-3-carboxylat

Lieferant: COMBI-BLOCKS

Synonyme: Ethyl 4-hydroxy-8-(trifluoromethyl)quinoline-3-carboxylate

HC-6355-5G HC-6355-1G
COBBHC-6355-5GEA 0 EUR
COBBHC-6355-5G COBBHC-6355-1G
Ethyl-4-hydroxy-8-(trifluormethyl)chinolin-3-carboxylat
Ethyl-4-hydroxy-8-(trifluormethyl)chinolin-3-carboxylat
Formel: C₁₃H₁₀F₃NO₃
MDL: MFCD09878378
CAS-Nummer: 23851-84-5
EINECS: 245-914-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR