Methyl-6,8-difluor-4-hydroxychinolin-2-carboxylat
Lieferant: COMBI-BLOCKS
HC-6951-5G
HC-6951-1G
COBBHC-6951-5GEA
884
EUR
COBBHC-6951-5G
COBBHC-6951-1G
Methyl-6,8-difluor-4-hydroxychinolin-2-carboxylat
Methyl-6,8-difluor-4-hydroxychinolin-2-carboxylat
Formel:
C₁₁H₇F₂NO₃ |
MDL:
MFCD11053876 CAS-Nummer: 1065074-53-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...