Ethyl-4-hydroxy-7,8-dimethylchinolin-3-carboxylat
Lieferant: COMBI-BLOCKS
HC-6553-5G
HC-6553-1G
COBBHC-6553-5GEA
375
EUR
COBBHC-6553-5G
COBBHC-6553-1G
Ethyl-4-hydroxy-7,8-dimethylchinolin-3-carboxylat
Ethyl-4-hydroxy-7,8-dimethylchinolin-3-carboxylat
Formel:
C₁₄H₁₅NO₃ |
MDL:
MFCD00665075 CAS-Nummer: 53164-33-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...