Methyl-8-methyl-4-hydroxy-5-(trifluormethyl)chinolin-2-carboxylat

Lieferant: COMBI-BLOCKS

HC-6856-5G HC-6856-1G
COBBHC-6856-5GEA 303 EUR
COBBHC-6856-5G COBBHC-6856-1G
Methyl-8-methyl-4-hydroxy-5-(trifluormethyl)chinolin-2-carboxylat
Methyl-8-methyl-4-hydroxy-5-(trifluormethyl)chinolin-2-carboxylat
Formel: C₁₃H₁₀F₃NO₃
MDL: MFCD12756446
CAS-Nummer: 1187386-20-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR