2-(2-Fluorphenyl)-4-chinolincarbonsäure
Lieferant: BIONET RESEARCH
3Y-0936.5
3Y-0936.1
3Y-0936.10
3Y-0936.500
BIOT3Y-0936.5EA
0
EUR
BIOT3Y-0936.5
BIOT3Y-0936.1
BIOT3Y-0936.10
BIOT3Y-0936.500
2-(2-Fluorphenyl)-4-chinolincarbonsäure
2-(2-Fluorophenyl)quinoline-4-carboxylic acid
Formel:
C₁₆H₁₀FNO₂ |
MDL:
MFCD06659551 CAS-Nummer: 1647-89-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...