4,4-Dimethylcyclohexen-1-ylboronsäurepinakolester
Lieferant: COMBI-BLOCKS
Synonyme:
2-(4,4-Dimethyl-1-cyclohexen-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PN-2526-5G
PN-2526-1G
COBBPN-2526-5GEA
812
EUR
COBBPN-2526-5G
COBBPN-2526-1G
4,4-Dimethylcyclohexen-1-ylboronsäurepinakolester
4,4-Dimethylcyclohexen-1-ylboronsäurepinakolester
Formel:
C₁₄H₂₅BO₂ Molecular Weight: 236,1581 g/mol |
MDL:
MFCD08690233 CAS-Nummer: 859217-67-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...