Methyl-3-isochinolincarboxylat

Lieferant: Thermo Scientific

BTB09392.1 BTB09392.10
MAYBBTB09392.1EA 0 EUR
MAYBBTB09392.1 MAYBBTB09392.10
Methyl-3-isochinolincarboxylat
Methyl-3-isochinolincarboxylat
Formel: C₁₁H₉NO₂
Molecular Weight: 187,2000 g/mol
MDL: MFCD00075138
CAS-Nummer: 27104-73-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR