2-(Allyloxy)-1-naphthaldehyd

Lieferant: BIONET RESEARCH

11F-907.500 11F-907.1 11F-907.5 11F-907.10
BIOT11F-907.500EA 0 EUR
BIOT11F-907.500 BIOT11F-907.1 BIOT11F-907.5 BIOT11F-907.10
2-(Allyloxy)-1-naphthaldehyd
2-(Allyloxy)-1-naphthaldehyd
Formel: C₁₄H₁₂O
MDL: MFCD00231645
CAS-Nummer: 19530-43-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR