2-(2-Propinyloxy)-1-naphthaldehyd
Lieferant: BIONET RESEARCH
7X-0711.500
7X-0711.1
7X-0711.5
7X-0711.10
BIOT7X-0711.500EA
0
EUR
BIOT7X-0711.500
BIOT7X-0711.1
BIOT7X-0711.5
BIOT7X-0711.10
2-(2-Propinyloxy)-1-naphthaldehyd
2-(2-Propinyloxy)-1-naphthaldehyd
Formel:
C₁₄H₁₀O |
MDL:
MFCD02629636 CAS-Nummer: 58758-48-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...