2-(Methylthio)naphthalin ≥98%
Lieferant: Thermo Fisher Scientific
B22368.03
B22368.06
B22368.03EA
92.7
EUR
B22368.03
B22368.06
2-(Methylthio)naphthalin ≥98%
2-(Methylthio)naphthalene
Purity: 98%
CAS No.: 7433-79-6
Molecular Formula: C11H10S
Formula Weight: 174.27
Odour: Stench
Melting Point: 62-63°
Beilstein Reference: 1859409
MDL No.: MFCD00051615
Formel:
C₁₁H₁₀S Schmelzpunkt: 62…63 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051615 CAS-Nummer: 7433-79-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...