2-(Methylthio)naphthalin

Lieferant: CHEMPUR

S11225.250 S11225.1
CHMPS11225.250EA 0 EUR
CHMPS11225.250 MAYBS11225.1
2-(Methylthio)naphthalin
2-(Methylthio)naphthalene
Formel: C₁₁H₁₀S
MDL: MFCD00051615
CAS-Nummer: 7433-79-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR