1-Naphthalinmethanol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Naphthalen-1-ylmethanol

184284-10G 184284-50G
SIAL184284-10GEA 71.5 EUR
SIAL184284-10G SIAL184284-50G
1-Naphthalinmethanol, Sigma-Aldrich®
1-Naphthalinmethanol
Formel: C₁₁H₁₀O
Molecular Weight: 158.20 g/mol
MDL: MFCD00004044
CAS-Nummer: 4780-79-4
EINECS: 225-324-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR