3-Methyl-2-naphthol
Lieferant: COMBI-BLOCKS
Synonyme:
2-Hydroxy-3-methylnaphthalene, 3-Methylnaphthalen-2-ol
OT-0588-5G
OT-0588-1G
COBBOT-0588-5GEA
1180
EUR
COBBOT-0588-5G
COBBOT-0588-1G
3-Methyl-2-naphthol
3-Methyl-2-naphthol
Formel:
C₁₁H₁₀O Molecular Weight: 158,1965 g/mol |
MDL:
MFCD09037754 CAS-Nummer: 17324-04-8 EINECS: 241-346-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...