3-(Hydroxymethyl)-2-hydroxynaphthalin

Lieferant: COMBI-BLOCKS

Synonyme: 3-(Hydroxymethyl)naphthalen-2-ol

OT-0723-1G OT-0723-5G
COBBOT-0723-1GEA 303 EUR
COBBOT-0723-1G COBBOT-0723-5G
3-(Hydroxymethyl)-2-hydroxynaphthalin
3-(Hydroxymethyl)-2-hydroxynaphthalin
Formel: C₁₁H₁₀O₂
Molecular Weight: 174.195 g/mol
MDL: MFCD11504950
CAS-Nummer: 30159-70-7

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR