2-(Benzyloxy)-1-naphthaldehyd
Lieferant: BIONET RESEARCH
4H-959.500
4H-959.1
4H-959.5
4H-959.10
BIOT4H-959.500EA
0
EUR
BIOT4H-959.500
BIOT4H-959.1
BIOT4H-959.5
BIOT4H-959.10
2-(Benzyloxy)-1-naphthaldehyd
2-(Benzyloxy)-1-naphthaldehyd
Formel:
C₁₈H₁₄O₂ |
MDL:
MFCD00202669 CAS-Nummer: 52805-48-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...