6-(Boc-methylamino)-3-pyridinboronsäure
Lieferant: COMBI-BLOCKS
Synonyme:
(6-((tert-Butoxycarbonyl)(methyl)amino)pyridin-3-yl)boronic acid
BB-8775-1G
BB-8775-250M
BB-8775-100M
COBBBB-8775-1GEA
1070
EUR
COBBBB-8775-1G
COBBBB-8775-250M
COBBBB-8775-100M
6-(Boc-methylamino)-3-pyridinboronsäure
6-(Boc-methylamino)-3-pyridinboronsäure
Formel:
C₁₁H₁₇BN₂O₄ Molecular Weight: 252.0800 g/mol |
MDL:
MFCD12756422 CAS-Nummer: 1218790-80-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...