Methyl-4-chlorpicolinat Hydrochlorid

Lieferant: COMBI-BLOCKS

CA-4100-5G CA-4100-25G
COBBCA-4100-5GEA 0 EUR
COBBCA-4100-5G COBBCA-4100-25G
Methyl-4-chlorpicolinat Hydrochlorid
Methyl-4-chlorpicolinat Hydrochlorid
Formel: C₇H₇Cl₂NO₂
MDL: MFCD05864729
CAS-Nummer: 176977-85-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR