6-(Boc-methylamino)-5-methyl-3-pyridinboronsäurepinakolester

Lieferant: COMBI-BLOCKS

Synonyme: 6-(N-Boc-methylamino)-5-methylpyridine-3-boronic acid pinacol ester

PN-8920-250M PN-8920-100M
COBBPN-8920-250MEA 310 EUR
COBBPN-8920-250M COBBPN-8920-100M
6-(Boc-methylamino)-5-methyl-3-pyridinboronsäurepinakolester
6-(Boc-methylamino)-5-methyl-3-pyridinboronsäurepinakolester
Formel: C₁₈H₂₉BN₂O₄
Molecular Weight: 348,25 g/mol
MDL: MFCD09037485
CAS-Nummer: 1032758-82-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR