Methyl-5-brom-3-methylpicolinat

Lieferant: COMBI-BLOCKS

Synonyme: 5-Bromo-3-methylpyridine-2-carboxylic acid methyl ester, Methyl 5-bromo-3-methylpyridine-2-carboxylate, Methyl 5-bromo-3-methylpicolinate

SS-3016-1G SS-3016-5G
COBBSS-3016-1GEA 102 EUR
COBBSS-3016-1G COBBSS-3016-5G
Methyl-5-brom-3-methylpicolinat
Methyl-5-brom-3-methylpicolinat
Formel: C₈H₈BrNO₃
Molecular Weight: 230.0586 g/mol
MDL: MFCD07375126
CAS-Nummer: 213771-32-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR