2-Formyl-3-picolin ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-Methylpyridine-2-carboxaldehyde, 3-Methylpicolinaldehyde
H56168.03
H56168.06
H56168.03EA
99.8
EUR
H56168.03
H56168.06
2-Formyl-3-picolin ≥97%
2-Formyl-3-picolin
Purity: 97%
CAS No.: 55589-47-4
Molecular Formula: C7H7NO
Formula Weight: 121.14
Flash Point: 73°(163°F)
Refractive Index: 1.538
MDL No.: MFCD02181144
Formel:
C₇H₇NO Molecular Weight: 121,14 g/mol Dichte: 1,08 Flammpunkt: 73 °C (163 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD02181144 CAS-Nummer: 55589-47-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...