2-Formyl-6-picolin
Lieferant: CHEMPUR
Synonyme:
6-Methylpyridine-2-carboxaldehyde, 6-Methyl-2-pyridinecarboxaldehyde
FL13661.POR
013661-25G
CHMPFL13661.POREA
0
EUR
CHMPFL13661.POR
FLUO013661-25G
2-Formyl-6-picolin
2-Formyl-6-picolin
Formel:
C₇H₇NO Molecular Weight: 121.14 g/mol |
MDL:
MFCD00006291 CAS-Nummer: 1122-72-1 EINECS: 214-359-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...