2-Methoxy-5-methyl-3-nitropyridin

Lieferant: COMBI-BLOCKS

PY-7062-1G PY-7062-5G
COBBPY-7062-1GEA 152 EUR
COBBPY-7062-1G COBBPY-7062-5G
2-Methoxy-5-methyl-3-nitropyridin
2-Methoxy-5-methyl-3-nitropyridine
Formel: C₇H₈N₂O₃
MDL: MFCD09258778
CAS-Nummer: 33252-62-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR