Ethyl-3-thiophencarboxylat ≥98%
Lieferant: Thermo Fisher Scientific
Synonyme:
Ethyl 3-Thiophenecarboxylate, 3-Thiophenecarboxylic acid ethyl ester
B22468.03
B22468.06
B22468.14
B22468.03EA
27.8
EUR
B22468.03
B22468.06
B22468.14
Ethyl-3-thiophencarboxylat ≥98%
Ethyl-3-thiophencarboxylat
Purity: 98%
CAS No.: 5751-80-4
Molecular Formula: C7H8O2S
Formula Weight: 156.20
Refractive Index: 1.5225
Boiling Point: 207-208°
MDL No.: MFCD02179226
Formel:
C₇H₈O₂S Siedepunkt: 207…208 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02179226 CAS-Nummer: 5751-80-4 EINECS: 227-275-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...