Methyl-3-methyl-2-thiophencarboxylat ≥99%
Lieferant: Thermo Fisher Scientific
Synonyme:
Methyl 3-methylthiophene-2-carboxylate
L18093.03
L18093.06
L18093.03EA
71.3
EUR
L18093.03
L18093.06
Methyl-3-methyl-2-thiophencarboxylat ≥99%
Methyl-3-methyl-2-thiophencarboxylat
Purity: 99%
CAS No.: 81452-54-2
Molecular Formula: C7H8O2S
Formula Weight: 156.20
Refractive Index: 1.5168
Boiling Point: 216-217°
MDL No.: MFCD00234332
Formel:
C₇H₈O₂S Molecular Weight: 156.20 g/mol Siedepunkt: 216…217 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00234332 CAS-Nummer: 81452-54-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...