4-(2-Cyanphenyl)morpholin

Lieferant: Thermo Scientific

Synonyme: 2-Morpholinobenzonitrile, 2-(Morpholin-4-yl)benzonitrile

SEW05816.1 SEW05816.10
MAYBSEW05816.1EA 0 EUR
MAYBSEW05816.1 MAYBSEW05816.10
4-(2-Cyanphenyl)morpholin
4-(2-Cyanphenyl)morpholin
Formel: C₁₁H₁₂N₂O
Molecular Weight: 188,2258 g/mol
MDL: MFCD01313390
CAS-Nummer: 204078-32-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR