5-Phenyl-1,3-cyclohexandion

Lieferant: MENAI

Synonyme: 5-Phenylcyclohexane-1,3-dione

ST199.25 ST199.50
MENAST199.25EA 0 EUR
MENAST199.25 MENAST199.50
5-Phenyl-1,3-cyclohexandion
5-Phenyl-1,3-cyclohexanedione
Formel: C₁₂H₁₂O₂
Molecular Weight: 188,23 g/mol
MDL: MFCD00051846
CAS-Nummer: 493-72-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR