Triethylbutylammoniumbromid ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(1-Butyl)triethylammonium bromide
B24128.14
B24128.22
B24128.14EA
80.3
EUR
B24128.14
B24128.22
Triethylbutylammoniumbromid ≥97%
Triethylbutylammoniumbromid
Purity: 97%
CAS No.: 13028-69-8
Molecular Formula: CH3(CH2)3N(CH2CH3)3Br
Formula Weight: 238.21
Melting Point: 212-215°
MDL No.: MFCD00055541
Formel:
C₁₀H₂₄BrN Molecular Weight: 238,21 g/mol Schmelzpunkt: 212…215 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00055541 CAS-Nummer: 13028-69-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...