3-(Aminomethyl)-5-brompyridin

Lieferant: COMBI-BLOCKS

OT-8104-1G OT-8104-250M
COBBOT-8104-1GEA 152 EUR
COBBOT-8104-1G COBBOT-8104-250M
3-(Aminomethyl)-5-brompyridin
3-(Aminomethyl)-5-bromopyridine
Formel: C₆H₇BrN₂
Molecular Weight: 187,0372 g/mol
MDL: MFCD06212876
CAS-Nummer: 135124-70-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR