4-Amino-2,3,5,6-tetrachlorpyridin

Lieferant: Thermo Scientific

Synonyme: 2,3,5,6-Tetrachloropyridin-4-amine

BTB14379.250 BTB14379.1
MAYBBTB14379.250EA 0 EUR
MAYBBTB14379.250 MAYBBTB14379.1
4-Amino-2,3,5,6-tetrachlorpyridin
4-Amino-2,3,5,6-tetrachloropyridine
Formel: C₅H₂Cl₄N₂
Molecular Weight: 231,8948 g/mol
MDL: MFCD00183239
CAS-Nummer: 2176-63-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR